Syringes for manual injection
Catalog # HEWL5190-1483
Supplier: Agilent
Specifications
- Rozchod:26
- Typ špičky:Bevelled
- Objem:10 µl
- Popis:Fixed needle
- Délka:50 mm
- Bal.:1
Specifications
About this item
Syringe with fixed needle with bevelled point. The plunger is made from a titanium/nickel alloy.
- Elastic, unbreakable plunger