ContactUsLinkComponent
Syringes for manual injection, plunger with PTFE tip for liquids and gases
Syringes for manual injection, plunger with PTFE tip for liquids and gases
Catalog # HEWL5190-1510
Supplier:  Agilent
Syringes for manual injection, plunger with PTFE tip for liquids and gases
Catalog # HEWL5190-1510
Supplier:  Agilent
Specifications
  • Rozchod:
    25
  • Objem:
    100 µl
  • Délka:
    50 mm
  • Bal.:
    1
Specifications
About this item
Syringes suitable for injecting liquids and gases. Fixed needle with bevelled point.
  • Replaceable plunger