Repeating dispensing system, PB600-1
Catalog # 549-0233
Supplier: HAMILTON BONADUZ
Specifications
- Popis:Repeating dispenser, PB600-1
- Bal.:1
Specifications
About this item
The PB600-1 works with Microliter™ or Gastight® syringes from 10 µl to 2,5 ml. Use the repeating dispenser with cemented needles (N), removable needles (RN), Luer tip (LT), or PTFE Luer lock (TLL) syringes.
- Up to 50 times with the push of a button
- Volumes are between 0,2 to 50 µl
Delivery: Supplied without syringe.