Disposable stirring spatulas
Disposable polystyrene spatulas/stirring rods, PS, 3 mm, 90 mm
Catalog # 441-0099
Supplier: KARTELL
Jump to:
Product Details & Documents
For various laboratory uses.
- Ideal as stirring rods for use in cuvettes
Documents
Specifications
- Ø:3 mm
- Popis:Disposable polystyrene spatulas/stirring rods
- Délka:90 mm
- Materiál:PS