Spoon, powder
170 mm, 40×10 mm
Katalogové číslo 231-1241
Dodavatel: RSG
Přejít na:
Product Details & Documents
Stainless steel 18/10. Multipurpose spoon for a wide range of uses.
- With cylindrical handle
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Documents
RSGA029.170_TDS_EN
Technický list
Specifikace
- Délka:170 mm
- Velikost lžíce:40×10 mm