Orders LinkContactUsLinkComponent
Spoons, POLY
 :   BOCHEM
Spoons, POLY
 :  BOCHEM
Spoons, POLY
Spoons, POLY
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
  • Sekvence
    Popis
    Délka
    Velikost lžíce
  • 01
    Straight
    155 mm
    40×20 mm
    1 * 1 KS
    1 * 1 KS - 144,00 Kč
    144,00 Kč
    1 * 1 KS
       144,00 Kč
    Specifications
    Cat. No.
    231-1068
  • 02
    Straight
    185 mm
    40×20 mm
    1 * 1 KS
    1 * 1 KS - 161,00 Kč
    161,00 Kč
    1 * 1 KS
       161,00 Kč
    Specifications
    Cat. No.
    231-1069
  • 03
    Sharply oblique
    190 mm
    25×20 mm
    1 * 1 KS
    1 * 1 KS - 130,00 Kč
    130,00 Kč
    1 * 1 KS
       130,00 Kč
    Specifications
    Cat. No.
    231-1261
  • 04
    Straight
    210 mm
    40×20 mm
    1 * 1 KS
    1 * 1 KS - 172,00 Kč
    172,00 Kč
    1 * 1 KS
       172,00 Kč
    Specifications
    Cat. No.
    231-1070
  • 05
    Oblique
    215 mm
    70×30 mm
    1 * 1 KS
    1 * 1 KS - 219,00 Kč
    219,00 Kč
    1 * 1 KS
       219,00 Kč
    Specifications
    Cat. No.
    231-1296
  • 06
    Oblique
    250 mm
    65×30 mm
    1 * 1 KS
    1 * 1 KS - 252,00 Kč
    252,00 Kč
    1 * 1 KS
       252,00 Kč
    Specifications
    Cat. No.
    231-0604
  • 07
    Straight
    255 mm
    40×20 mm
    1 * 1 KS
    1 * 1 KS - 188,00 Kč
    188,00 Kč
    1 * 1 KS
       188,00 Kč
    Specifications
    Cat. No.
    231-1269
  • 08
    Straight
    300 mm
    40×20 mm
    1 * 1 KS
    1 * 1 KS - 200,00 Kč
    200,00 Kč
    1 * 1 KS
       200,00 Kč
    Specifications
    Cat. No.
    231-1071
  • 09
    Oblique
    300 mm
    65×28 mm
    1 * 1 KS
    1 * 1 KS - 281,00 Kč
    281,00 Kč
    1 * 1 KS
       281,00 Kč
    Specifications
    Cat. No.
    231-1297
  • 10
    Oblique
    395 mm
    65×30 mm
    1 * 1 KS
    1 * 1 KS - 370,00 Kč
    370,00 Kč
    1 * 1 KS
       370,00 Kč
    Specifications
    Cat. No.
    231-1270
  • 11
    Oblique
    500 mm
    65×30 mm
    1 * 1 KS
    1 * 1 KS - 410,00 Kč
    410,00 Kč
    1 * 1 KS
       410,00 Kč
    Specifications
    Cat. No.
    231-0605