Orders LinkContactUsLinkComponent
Spatula set
Spatula set
Set of 6 spatulas and 1 pair of tweezers
Katalogové číslo 231-2176
Dodavatel:   RSG
Spatula set
Katalogové číslo 231-2176
Dodavatel:  RSG
Spatula set
Spatula set
Set of 6 spatulas and 1 pair of tweezers
Přejít na:
Price
2 160,00 Kč
Přihlásit se pro dostupnost
1 * 1 SET
Product Details & Documents
Stainless steel 18/10. Useful combination of double spatulas, spoon spatulas and forceps. Universal usage in laboratories and industry.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Comes in practical imitation leather case
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as tweezers (straight, blunt, 145 mm long).
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as forceps (straight, blunt, 145 mm long).
Specifikace
  • Popis:
    Set of 6 spatulas and 1 pair of tweezers