Capping tool, Cap-It
Catalog # 732-4817
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Capping tool, Cap-It
- Bal.:1
Specifications
About this item
Designed to assist users in applying domed caps to tube strips and plates.
- Minimises the pressure required to seal the caps effectively
- Enables multiple tubes to be sealed simultaneously