Sealing tape with dispenser, Nunc™
Catalog # 391-0113
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Sealing tape with dispenser
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:1.000
Specifications
About this item
Polyester tape with acrylic adhesive. Easy peel, easy seal general-purpose tape with a faster application time than normal tape in loose sheets.
- Temperature stable from −20 to +120 °C
- Non-pierceable by pipette
Colour: clear
External L×W: 121×78 mm (plus handle 9×78 mm)