Sealing tape for MicroWell™ plates, Nunc™
Katalogové číslo 732-2610
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Sealing tape, polyester
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:200
Specifications
About this item
Polyester film with acrylic adhesive. Provides an effective seal for all MicroWell™ plates used in PCR and cell culture.
- Resistant to temperatures between −70 and +100 °C
- Minimum evaporation and well-to-well cross-contamination
Dimensions: 134×80 mm
Dimensions: 134×80 mm