Universal scissors, bent
180 mm
Katalogové číslo 233-1152
Dodavatel: HAMMACHER KARL
Přejít na:
Product Details & Documents
Stainless steel, with plastic handles.
- Useful for many applications, perfect for cutting cardboard and metal foils
- Corrosion resistant, autoclavable
- Easy action
Documents
HAMMHSB601-18_TDS_EN
Technický list
Specifikace
- Délka:180 mm