Physiology scissors, straight
Dodavatel: HAMMACHER KARL
Přejít na:
O tomto produktu
Stainless steel 18/10, anti-magnetic.
- Sharp-sharp
- Ideal for a wide range of laboratory applications
- Acid-resistant
- Autoclavable
2 Dostupných variant níže
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- Sharp-sharp
- Ideal for a wide range of laboratory applications
- Acid-resistant
- Autoclavable
Product Family Options
Informace o produktu
- TypeDélkaDostupnostPrice