Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, 115 mm
Catalog # 233-2124
Supplier:   BOCHEM
Microscopy scissors
Catalog # 233-2124
Supplier:  BOCHEM
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, 115 mm
Price
174,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Corrosion resistant
  • Autoclavable
  • Very sharp tips
Specifications
  • Type:
    Straight, sharp/sharp
  • Délka:
    115 mm