Racks for microcentrifuge tubes
HEATHS4284A
: HEATHROW
- Pro zkumavky:1,5/2,0 ml
- Poč. otvorů:20
- Barva:Modrá
- Š×H×V:212×89×51 mm
- Bal.:6
PP, autoclavable.
- Racks lock together on each side permitting the user to attach several for larger experiments
- “Hold-it” finger slot on the front
- Internal ribbing for stability