0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Podnosy/Misky/Kelímky
Podnosy na pipety
Pipette tray for drawer
Pipette tray for drawer
612-1805
:
KARTELL
Pipette tray for drawer
612-1805
:
KARTELL
Popis:
Pipette tray for drawer
Bal.:
1
PVC, white.
Designed to hold about 30 pipettes
Four compartments to accommodate 1, 2, 5, 10 ml pipettes
W×D×H: 425×300×30 mm.