Držáky svorek na pipety, Clamp-ette®
Katalogové číslo 613-3900
Dodavatel: HEATHROW
Specifikace
- Určeno pro:3 pipettes
- Barva:Modrá
- Popis:Pipettor clamp
- Sekvence:02
- Bal.:1
Specifikace
O tomto produktu
PP, coloured.
- Stable, sturdy construction
- For one to three pipettes of the most widely used brands
- Secures to tabletops, benches and shelves with a twist of the fingertips
- Autoclavable