Pipette stand, rotary, SP Bel-Art
Catalog # 612-0013
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø×H:178×178 mm
- Barva:Bílá
- Bal.:1
Specifications
About this item
Pipette support stand made of white PP.
- For 28 pipettes
- 10 holes, 14 mm Ø
- 18 holes, 11 mm Ø
Ø×H: 178×178 mm