Orders LinkContactUsLinkComponent
Pipette stand, rotary
 :   KARTELL
Pipette stand, rotary
 :  KARTELL
Pipette stand, rotary
Pipette stand, rotary
PP, with circular PVC base (both references to be ordered together to have the complete system).
  • For 18 pipettes Ø <10 mm and 10 pipettes Ø <15 mm
  • The base on which the pipettes rest is provided with small drainage holes
  • With fixing ring for height adjustment on shaft of circular base
Product Family Options
  • Popis
    Ø×H
  • Cicular base with PVC rod
    230×460 mm
    1 * 1 KS
    1 * 1 KS - 605,00 Kč
    605,00 Kč
    1 * 1 KS
       605,00 Kč
  • Pipette stand without base
    180×180 mm
    1 * 1 KS
    1 * 1 KS - 955,00 Kč
    955,00 Kč
    1 * 1 KS
       955,00 Kč