Bulb pipettes with syringe, one mark
Catalog # 612-1347
Supplier: HIRSCHMANN
Specifications
- Kapacita:50 ml
- Bal.:1
- Tolerance:0,05 ± ml
Specifications
About this item
Glass.
- Silver diffusion ink, durable brown graduations
- Attached syringe for simple, risk-free pipetting