Orders LinkContactUsLinkComponent
Mops, LoopMop™
 :   MICRONOVA
Mops, LoopMop™
 :  MICRONOVA
Mops, LoopMop™
Mops, LoopMop™
Knitted polyester mop for applying disinfectant in pharmaceutical, aseptic processing and medical device applications. Absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. Mop heads can be used directly with the fibreglass/polypropylene handle or attached to the universal handle via the quick disconnect adapter.
  • Can be laundered or autoclaved
  • Low linting
  • No raw edges
 : All components must be ordered separately.
  • Catalog No:
  • 129-0241
  • MICRMZLM5-12IR
  • Environmentally Preferable:
  • Waste Reducing
Product Family Options
  • Popis
  • LoopMop™
    1 * 1 KS
    1 * 1 KS - 1 470,00 Kč
    1 470,00 Kč
    1 * 1 KS
       1 470,00 Kč
    Specifications
    Cat. No.
    129-0241
    Environmentally Preferable
  • LoopMop™, irradiated
    1 * 24 KS
    1 * 24 KS - 40 460,00 Kč
    40 460,00 Kč
    1 * 24 KS
       40 460,00 Kč
      Waste Reducing
    Specifications
    Cat. No.
    MICRMZLM5-12IR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Accessories
  •  
    Sekvence
    Popis
    Délka
  • 129-0551
    01
    Prodlužovací nerezová tyč
    965 - 1829 mm
    1 * 1 KS - 7 840,00 Kč
  •  
    Sekvence
    Popis
    Délka
  • 129-0062
    09
    Standardní nerezová tyč
    155 cm
    1 * 1 KS - 3 600,00 Kč
  •  
    Sekvence
    Popis
    Délka
  • 129-0176
    08
    Stainless steel quick disconnect handle
    1295 cm
    1 * 1 KS - 3 490,00 Kč
  •  
    Sekvence
    Popis
  • 129-0170
    04
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    1 * 1 KS - 3 430,00 Kč