Adhesion slides, Menzel Gläser, Polysine®
Catalog # 631-1349
Supplier: Epredia
Specifications
- Barva:Bílá
- Popis:Adhesion slides
- Bal.:72
Specifications
About this item
These slides, made using a technology that electrostatically and chemically attracts fresh cell preparations, formalin-fixed Bouin's or alcohol-fixed paraffin-embedded tissues.
- 90° ground edges with frosted ends for labelling
- Adhesion superior to glue-, protein- or silane-treated slides
- Thick tissue sections of 2 to 5 µm
Unaffected by enzyme pre-digestion or heating, tissue loss is minimised in prolonged in-situ DNA hybridisation techniques and in immunocytochemical methods.