Workstation and stacking system for PCR tubes and plates
Katalogové číslo 732-4818
Dodavatel: Thermo Fisher Scientific
Specifikace
- Poč. otvorů:96/48
- Popis:Workstation and stacking system
- Bal.:10
Specifikace
O tomto produktu
Convenient platform for tube handling and reagent dispensing before and after sample processing.
- Multiple workstations can be securely stacked
- Can be stored at down to −190 °C
Holds:
96×0,2 ml V-bottom tubes
96×0,3 ml U-bottom Omnistrips
Thermo-Tube plates
Thermo-Fast® non-skirted 96-well plates
Thermo-Fast® semi-skirted 96-well plates
48×0,5 ml Thermo-Tubes