PCR plates, 384-well, Thermo-Fast® 384 Mark II
Katalogové číslo 732-4912
Dodavatel: Thermo Fisher Scientific
Specifikace
- Poč. jamek:384
- Profil:Standard
- Lem:Fully skirted
- Well colour:Průhledná
- Bal.:100
- Objem jamky:0,04 ml
Specifikace
O tomto produktu
Moulded from ultra low retention PP that assists in preventing adhesion of samples to the side walls. Due to the increased well volume, post-PCR applications such as enzymatic digestions and ethanol precipitation can be carried out in the wells. Compatible with all leading 384 block thermal cyclers.
- Fully skirted for compatibility with automated systems
- 8 holes in the skirt aid plate positioning and removal from the thermal cycler block
- Cut away corner aids orientation
- Certified free from DNase, RNase and human genomic DNA
Maximum well volume: 55 µl
Working volume: 40 µl