PCR plates, 384-well, Thermo-Fast®
Catalog # 732-4961
Supplier: Thermo Fisher Scientific
Specifications
- Poč. jamek:384
- Lem:Fully skirted
- Well colour:Průhledná
- Bal.:50
Specifications
About this item
PP. Designed for high throughput PCR, this 384-well plate is compatible with the majority of 384 block PCR, qPCR and sequencing instruments.
- The full skirt and rigid plate deck minimise distortion during PCR making it suitable for use with robotic systems
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
- Certified free from DNase, RNase and human genomic DNA
- Alphanumeric grid referencing and cut away corners aid orientation
Maximum well volume: 40 µl
Working volume: 25 µl