PCR plates, 96-well, rigid, semi-skirted, Thermo-Fast®
Catalog # 732-4924
Supplier: Thermo Fisher Scientific
Specifications
- Poč. jamek:96
- Profil:Standard
- Lem:Semi-skirted
- Well colour:Průhledná
- Bal.:25
- Objem jamky:0,2 ml
Specifications
About this item
PP. A rigid version of the standard semi-skirted plate for applications where extra plate rigidity is required.
Colour: Clear