CovaLink™ NH modules, Nunc™
Catalog # 735-0153
Supplier: Thermo Fisher Scientific
Specifications
- Barva:Průhledná
- Popis:F8×12 module, clear, framed
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Bal.:30
Specifications
About this item
PS. Developed to bind compounds covalently to plastic surfaces. For use with molecules which do not adsorb easily to a traditional surface. Framed modules with alphanumeric orientation (external dimensions: 128×86 mm) fit all standard equipment.
- Very thorough washing possible with high ionic strength solutions
- Minimum non-specific binding
- Allows site-specific binding via, for example, a carboxylic group of peptide or phosphate group in the 5' end of nucleic acids
- Free-standing modules with flat (F) wells
Recommended working volume: 100 µl
Recommended working volume: 100 µl