Microplates, 384-well
Product Details & Documents
Ideal for high throughput screening, these 384-well microplates allow miniaturisation of assays and many other applications, including high volume DNA library manipulation and solid phase binding assays. Available with a choice of clear microplates for colorimetric applications, white plates for luminometric/fluorometric applications, and black microplates for fluorometric applications. There are two different designs, both of which incorporate flat bottom wells and a standard microplate footprint. The first type has a square well shape with rounded corners for an increased well volume. It has the standard microplate height and a total well volume of 120 µl. The second type has round wells and a low profile, with a total well volume of 70 µl.
- Flat bottom wells for optimum optical characteristics
- Standard footprint compatible with automated handling equipment
- Choice of rounded square or round well shape
Specifikace
- Poč. jamek:384
- Sterilní:-
- Barva:Průhledná
- Popis:384-well plate
- Cat. No.:735-0577
- Certifikát ochrany životního prostředí:ISO 14001 (Certificate Registration No. EMS 623400)
- Environmentally Preferable:Low Manufacturing Impact