Orders LinkContactUsLinkComponent
Stools, mobile, metal
Stools, mobile, metal
Supplier:   WEDO WERNER DORSCH
Stools, mobile, metal
Supplier:  WEDO WERNER DORSCH
Certificates
Stools, mobile, metal
Stools, mobile, metal
4 Options Available Below
Product Details & Documents
Manufactured from strong powder coated steel and mounted on three castors allowing complete mobility. When in use the castors retract, providing a secure platform.
  • Non slip rubber mats, lead- and cadmium-free lacquer
  • TÜV GS approved, supports up to 150 kg
  • Snap-lock assembly, no tools required
EN 14183 (DIN 4569)
Certifications: EN 14183 (DIN 4569).
Product Family Options
Product Information
  • Popis
    Barva
    Výška
    Ø horní části / Ø dna (mm)
    Hmotnost
    Availability
    Price
  • Kick stool, metal
    Modrá
    440 mm
    295/435 mm
    5,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 600,00 Kč
    3 600,00 Kč
    1 * 1 KS
       3 600,00 Kč
  • Kick stool, metal
    Světle šedá
    440 mm
    295/435 mm
    5,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 600,00 Kč
    3 600,00 Kč
    1 * 1 KS
       3 600,00 Kč
  • Kick stool, metal
    Černá
    440 mm
    295/435 mm
    5,7 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS
  • Kick stool, metal
    Červený
    440 mm
    295/435 mm
    5,7 kg