Footrest, Swing
Footrest, swing, Černá, 4770×320×105 mm, 1,7 kg
Catalog # 139-9805
Supplier: WEDO WERNER DORSCH
Jump to:
Product Details & Documents
A stylish footrest that is fully adjustable from a normal seated position. Ideal for use at shared workstations.
- Ribbed platform provides extra grip
Documents
VWR® For Your Space
Product Literature
Specifications
- Hmotnost:1,7 kg
- Barva:Černá
- Popis:Footrest, swing
- Š×H×V:4770×320×105 mm