Radioactivity indicator dots, SP Bel-Art
116-1095
- Barva:Červená/žlutá
- Popis:Indicator dots
- Rozměry:6 mm
- Balené:1000 labels/roll
- Bal.:1 Roll
Stick-on dots with the radiation symbol on yellow background, which enables easy labelling of radioactive vessels.
- Ideal for marking micro-centrifuge tubes
Colour coding complies with OSHA regulations.