Staining jars
Staining jar, Schiefferdecker type, 85×70×50 mm
Katalogové číslo 631-9131
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
Made from PMP (TPX).
This rectangular jar is supplied with a lid and can hold 10 slides or 20 slides back-to-back.
Specifikace
- Popis:Staining jar, Schiefferdecker type
- Š×H×V:85×70×50 mm