Orders LinkContactUsLinkComponent
Staining jars acc. to Hellendahl
Staining jars acc. to Hellendahl
Staining jar, Hellendahl type, 57×57×90 mm
Katalogové číslo 631-9130
Dodavatel:   KARTELL
Staining jars acc. to Hellendahl
Katalogové číslo 631-9130
Dodavatel:  KARTELL
Staining jars acc. to Hellendahl
Staining jars acc. to Hellendahl
Staining jar, Hellendahl type, 57×57×90 mm
Přejít na:
Price
366,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PMP (TPX), transparent.
  • Extension and cover
Staining jar similar to Hellendahl design for 16 standard microscope slides 76×26 mm (in pairs, back to back).
Specifikace
  • Popis:
    Staining jar, Hellendahl type
  • Š×H×V:
    57×57×90 mm