Orders LinkContactUsLinkComponent
Staining jars acc. to Coplin, WHEATON®
Staining jars acc. to Coplin, WHEATON®
Staining jar acc. to Coplin, with screw cap, 26×26×70 mm
Katalogové číslo 720-0707
Dodavatel:   DWK Life Sciences
Staining jars acc. to Coplin, WHEATON®
Katalogové číslo 720-0707
Dodavatel:  DWK Life Sciences
Staining jars acc. to Coplin, WHEATON®
Staining jars acc. to Coplin, WHEATON®
Staining jar acc. to Coplin, with screw cap, 26×26×70 mm
Price
5 290,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
Soda-lime glass.
  • Slides extend above the opening
  • Easy manipulation without using forceps
  • Stable rectangular base
This staining jar has the capacity for up to 10 standard slides of 76×26 mm with white PP screw cap.
Specifikace
  • Popis:
    Staining jar acc. to Coplin, with screw cap
  • Š×H×V:
    26×26×70 mm
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Slide grip for Wheaton Coplin staining jars
    Slide grip for Wheaton Coplin staining jars
    720-0709
    Přihlásit se pro dostupnost
    1 * 2 KS - 1 480,00 Kč