Staining jars acc. to Hellendahl, WHEATON®
720-0710
- Popis:Staining jar acc. to Hellendahl
- Š×H×V:42×26×85 mm
- Bal.:6
Soda-lime glass.
- Easy to use due to wide opening
- For staining or use as a TLC developing chamber
Staining jar holds eight individual 76×26 mm standard slides or 16 slides back to back.