Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Staining jar with 2 lids, 81×101×70 mm
Katalogové číslo 631-9511
Dodavatel:   KARTELL
Staining jars
Katalogové číslo 631-9511
Dodavatel:  KARTELL
Staining jars
Staining jars
Staining jar with 2 lids, 81×101×70 mm
Přejít na:
Price
677,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PMP.
  • One lid for when rack is not in use, providing evaporation free storage of staining liquid
  • Second lid with special opening to allow handle of staining rack (631-9520) to be inserted
Staining jar supplied with two lids.
Informace o dodávce: For use with 631-9520.
Upozornení: Xylene can cause etching after two days at room temperature.
Specifikace
  • Popis:
    Staining jar with 2 lids
  • Š×H×V:
    81×101×70 mm