Forceps for cover glasses
Catalog # 232-0111
Supplier: USBECK Laborgeräte
Specifications
- Type:Forceps for cover glasses, curved
- Délka:105 mm
- Sekvence:01
- Bal.:1
Specifications
About this item
Steel, nickel-plated.
- For easy, secure handling of cover glasses
- Can also be used for other laboratory equipment, such as filter discs
- Easy to clean