Orders LinkContactUsLinkComponent
Forceps
 :   VITLAB
Forceps
 :  VITLAB
Forceps
Forceps
PMP, white. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance
  • Autoclavable
  • Robust, pliable, self-sprung
Product Family Options
  • Type
    Délka
  • Straight, pointed
    115 mm
    1 * 1 KS
    1 * 1 KS - 44,00 Kč
    44,00 Kč
    1 * 1 KS
       44,00 Kč
  • Straight, pointed
    145 mm
    1 * 1 KS
    1 * 1 KS - 53,50 Kč
    53,50 Kč
    1 * 1 KS
       53,50 Kč