Orders LinkContactUsLinkComponent
Forceps, precision, with locking clip
Forceps, precision, with locking clip
Forceps, precision, with locking clip
Supplier:  OUTILS RUBIS
Certificates
Forceps, precision, with locking clip
Forceps, precision, with locking clip
2 Options Available Below
Product Details & Documents
Stainless steel. Forceps with very precise, fine tips. For applications that require high precision such as in microscopy, biology and microbiology.
  • Acid-resistant, resistant to hydrofluoric acid and nitric acid
  • 100% antimagnetic
  • Extra fine tips
Documents
Certificates
Search by Lot Number
    Images 360
  • 5L
Product Family Options
Product Information
  • Popis
    Type
    Délka
    Availability
    Price
  • Straight, ultra fine tips
    3L, with locking clip
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 1 180,00 Kč
    1 180,00 Kč
    1 * 1 KS
       1 180,00 Kč
  • Straight, ultra fine tips (tower tip)
    5L, with locking clip
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 1 030,00 Kč
    1 030,00 Kč
    1 * 1 KS
       1 030,00 Kč