Forceps
105 mm
Katalogové číslo 232-0039
Dodavatel: MURRAY S
Přejít na:
Product Details & Documents
Steel, nickel plated
- With flat, curved ends
- Robust with easy action
- Very good thermal resistance, good chemical resistance
- Light and easy to clean
Ideal for cover glasses, slides and filter discs.
Specifikace
- Délka:105 mm