Flow indicator paddle wheels, Roto-Flo™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PS paddle wheels for flow visualisation of liquids or gases.
- Internal flow channel of 5 mm
- Paddle wheel shell of 25 mm
- Overall length of 51 mm
- Maximum recommended pressure of 10 psi
Minimum flow of 0,5 liters per minute − Maximum flow of 10 liters per minute.
Caution: Not for use with corrosive chemicals or fluids with a temperature in excess of 64 °C (150 °F).
Specifications
- Catalog No:
- 618-1140
- 618-1142
- 618-1141
- Připojení:
- Barbed
- Barbed
- Barbed