Flasks, iodine with collar and iodine stopper
Catalog # 201-1162
Dodavatel: Lenz Laborglas GmbH & CO.KG
Specifikace
- Ø vněj.:85 mm
- Velikost objímky:29/32 NS
- Kapacita:250 ml
- Bal.:1
Specifikace
O tomto produktu
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.