O tomto produktu
These filter papers are made of 100% cotton linters. They are guaranteed to be free of any residual acids and they have an extremely low percentage of ash content. As a result, these filters are virtually ash-free. All Sartorius ashless filter papers are characterised by high wet strength and high purity and are therefore especially suitable for quantitative analysis, Büchner funnels, pressure or vacuum filtration.
388: 'Black dot filter' For coarse-flocculent and voluminous precipitates, especially iron, aluminium and chromium hydroxides; copper, bismuth, cobalt and iron sulphides; various organic, coloured metal deposits; determination of silicon in analysis of steel and pig iron; for acidic and slightly alkaline solutions; pressure filtration.
389: 'White dot filter' The most used grade for analyses; for relatively coarse precipitates such as silver, arsenic, antimony, cadmium, lead, iron and manganese sulphides; lead chromate; ammonium phosphorous molybdate; alkaline earth carbonates '...' for acidic and slightly alkaline solutions; pressure filtration.
390: 'Green dot filter' Filtration of fine precipitates; barium sulphate; lead molybdate; lead dioxide; calcium hydroxide; calcium fluoride; nickel, zinc and stannic sulphides; for acidic and slightly alkaline solutions; pressure filtration.
393: 'Purple dot filter' Ideal for particularly difficult filtration conditions and especially fine-grained precipitates; barium sulphate; copper oxide.
Specifikace
- Catalog No:
- 516-7044
- SARTFT-2-103580580
- 515-1202
- 516-7042
- SARTFT-2-102580580
- 515-1146
- SARTFT-2-101580580
- SARTFT-2-127580580
- 515-1162
- 515-1226
- 515-1184
- 515-1183
- 515-1143
- 515-1208
- 515-1203
- Třída:
- 393
- 390
- 391
- 393
- 389
- 388
- 388
- 393
- 389
- 392
- 390
- 390
- 388
- 391
- 391
- Hmotnost:
- 100 g/m²
- 84 g/m²
- 84 g/m²
- 100 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 100 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- Záchyt částic:
- 1 - 2 µm
- 3 - 5 µm
- 2 - 3 µm
- 1 - 2 µm
- 8 - 12 µm
- 12 - 15 µm
- 12 - 15 µm
- 1 µm
- 8 - 12 µm
- 5 - 8 µm
- 3 - 5 µm
- 3 - 5 µm
- 12 - 15 µm
- 2 - 3 µm
- 2 - 3 µm
- Rychlost filtrace:
- 300 s
- 100 s
- 180 s
- 300 s
- 20 s
- 10 s
- 10 s
- 300 s
- 20 s
- 50 s
- 100 s
- 100 s
- 10 s
- 180 s
- 180 s
- Obsah popela:
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%
- ≤0,010%