Multi-compartment dispenser
Multi-compartment dispenser, acrylic, Průhledná, 508×305×457 mm
Katalogové číslo 113-0032
Dodavatel: AK
Přejít na:
Product Details & Documents
Acrylic dispenser for the storage of gloves, caps, hairnets, beard covers, shoe covers.
- One large and two small compartments
- Easy loading
- Top lid access
Specifikace
- Barva:Průhledná
- Popis:Multi-compartment dispenser, acrylic
- Š×H×V:508×305×457 mm
- Cat. No.:113-0032
- Typ dávkovače:Floor-standing/wall-mountable
- Materiál:Akryl
- PPE type:Multi-use