Orders LinkContactUsLinkComponent
Finger cot dispenser
Finger cot dispenser
Finger cot dispenser, Bílá, 152×216×318 mm
Catalog # 113-0030
Supplier:   AK
Finger cot dispenser
Catalog # 113-0030
Supplier:  AK
Finger cot dispenser
Finger cot dispenser
Finger cot dispenser, Bílá, 152×216×318 mm
Price
6 370,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
This finger cot dispenser is suitable for use in cleanrooms.
  • Easy loading
  • Top lid access
  • Wall-mounted or free-standing
Specifications
  • Barva:
    Bílá
  • Popis:
    Finger cot dispenser
  • Š×H×V:
    152×216×318 mm