Desiccator plates, Nalgene®
467-3100
- Ø:230 mm
- Type:Ceramic metal composite
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:1
Unbreakable desiccator plates with fire polished, corrosion resistant, inert, non stick glass surface bonded to metal. Suitable for use with desiccators: 467-3120, 467-3110 and 467-3112.
- Greater resistance to thermal shock than porcelain
- Marked with numbered quadrants for easy reference
- Autoclavable