Orders LinkContactUsLinkComponent
Sample container with snap-on cap
Sample container with snap-on cap
Supplier:   VITLAB
Sample container with snap-on cap
Supplier:  VITLAB
Certificates
Sample container with snap-on cap
Sample container with snap-on cap
4 Options Available Below
Product Details & Documents
PP, transparent with clear PE-LD snap on cap.
  • Available in four sizes
  • Leakproof snap-on cap
Ideal for collecting, storing and transporting samples.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Product Family Options
Product Information
  • Kapacita
    Ø vněj.
    Výška
    Availability
    Price
  • 5 ml
    20 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 25 KS
    1 * 25 KS - 832,00 Kč
    832,00 Kč
    1 * 25 KS
       832,00 Kč
  • 18 ml
    22 mm
    57 mm
    Log in for availability
    Log in for availability
    1 * 25 KS
    1 * 25 KS - 1 050,00 Kč
    1 050,00 Kč
    1 * 25 KS
       1 050,00 Kč
  • 50 ml
    30 mm
    97 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 550,00 Kč
    550,00 Kč
    1 * 10 KS
       550,00 Kč
  • 160 ml
    50 mm
    110 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 643,00 Kč
    643,00 Kč
    1 * 10 KS
       643,00 Kč