Orders LinkContactUsLinkComponent
Beard covers, PP
Bílá
  113-0693
Beard covers, PP
  113-0693
Beard covers, PP
Beard covers, PP
Bílá
3 420,00 Kč
1 * 1.000 KS
Comfortable beard cover made from non woven PP, for non critical applications.
  • With elastic straps
  • With welded seams
Weight: 8 g
 : Dispenser box of 1000.
  • Barva:
    Bílá
  • Hmotnost:
    8 g
  • Cat. No.:
    113-0693