Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Cell scrapers
Supplier:  Greiner Bio-One
Certificates
Cell scrapers
2 Options Available Below
Product Details & Documents
Designed to facilitate the gentle mechanical removal of adherent cells from growth surfaces. Each scraper has a 18 mm pivoted (pivot angle 60°) scraper blade, which allows access to poorly accessible surfaces.
  • Optimised blade design for maximum cell harvest
  • Uniform contact with the growth surface, minimal mechanical strain and efficient cell harvest, even from poorly accessible surfaces
  • Minimises any accumulation of the cell suspension to the blade structure
Packaging: Individually packaged.
Documents
Product Family Options
Product Information
  • Popis
    Celková délka
    Availability
    Price
  • Cell scraper with 18 mm pivoted blade
    280 mm
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 4 230,00 Kč
    4 230,00 Kč
    1 * 100 KS
       4 230,00 Kč
  • Cell scraper with 18 mm pivoted blade
    400 mm
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 10 240,00 Kč
    10 240,00 Kč
    1 * 100 KS
       10 240,00 Kč