SP Bel-Art 'The Collector™' Blood Collecting Tray Set, Bel-Art Products, a part of SP
Catalog # 129-1015
Supplier: Bel-Art Products, a Part of SP
Specifications
- Popis:"The Collector™"
- Š×H×V:356×292×51 mm
- Bal.:1
Specifications
About this item
PP, white.
- With two 40-place test tube racks (tubes up to 16 mm Ext.Ø.)
- With one 18-place slide holder
- With large requisition section
- Adjustable partitions for syringes, needles, cotton etc.
"The Collector" is a versatile carrying tray particularly suited for use in clinical laboratories. The tray will not sag even when fully loaded. It is steam autoclavable at 121 °C. The handle is an end-mounted, detachable loop (178 mm high).