Byretová samonulovací sada, Nalgene®
Catalog # 612-2587
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Self-Zero Kit
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:1
Specifications
About this item
Makes any plastic or glass burette self-zeroing.
Consists of a 1000 ml LDPE bottle, 1,2 m clear PVC tubing, PP fill tube and adapter.